Order Entry
United States
ContactUsLinkComponent
 
Transgenic Dough Diet™ for Rodents, Sterile, Bio-Serv
  89067-442
 :  Bio-Serv
undefined
Transgenic Dough Diet™ for Rodents, Sterile, Bio-Serv
  89067-442
 :  Bio-Serv
 :  S3472

 

  • Description:
    Transgenic Dough, Bacon Flavor
  • Environmentally Preferable:
  • Cat. No.:
    89067-442
  • Packaging:
    1 kg/Bag
  • Supplier no.:
    S3472

 

 

Nutritionally complete, soft diet with higher protein and fat levels to comply with NRC and Transgenic-users' recommendations

  • Designed for rodents with chewing, dental or mobility impairments
  • Cookie dough consistency replaces labor intensive, nutrient-diluted mush diets
  • Ideal for recovering rodents and weanlings or any time a soft diet is needed
  • Rich bacon flavor stimulates appetite in dogs and rodents
  • Place in feeder cup or on cage floor
  • Sterilized by gamma irradiation
  • Nutritionally assayed
  • Shelf Life: 6 months refrigerated, 12 months frozen

Ingredients: Sucrose, Whole Milk Powder, Casein, Soybean Oil, Cellulose, Corn Syrup, Vitamin Mix, Mineral Mix, Glycerin, Bacon Flavor, DL-Methionine, Choline Bitartrate, Phenylalanine, tBHQ.

Caloric value: 3.83 kcal/g
Nutritional ash content: 3.3%
Nutritional carbohydrate content: 46.5%
Nutritional fat content: 12.4%
Nutritional fibre content: 4.8%
Nutritional moisture content: <12%
Nutritional protein content: 21.2%