Order Entry
United States
ContactUsLinkComponent
Rodent BLT's™ Diet, Bio-Serv
Rodent BLT's™ Diet, Bio-Serv
Catalog # 89067-428
Supplier:  Bio-Serv
undefined
Rodent BLT's™ Diet, Bio-Serv
Catalog # 89067-428
Supplier:  Bio-Serv
Supplier Number:  F05072

Specifications

  • Ø:
    2.54 cm (1")
  • Description:
    Rodent BLT's, Bacon Flavor
  • Height:
    0.64 cm (¼")
  • Size:
    5 g wafers
  • Environmentally Preferable:
  • Cat. No.:
    89067-428
  • Packaging:
    1,000/Box
  • Supplier no.:
    F05072

Specifications

About this item

Nutritionally complete, 5 g grain-based bacon-flavored diet

  • Feed as an appetite stimulant, treat or sole source diet
  • Excellent for adding medications and test articles
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.

Caloric value: 3.39 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 4.3%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%