Specifications
- Ø:2.54 cm (1")
- Description:Nutty Monkey Treats
- Height:0.64 cm (¼")
- Size:5 g wafers
- Cat. No.:89067-324
- Packaging:1,000/Box
- Supplier no.:F3788
Specifications
About this item
Nutritionally complete grain-based diet with bits of peanuts
- Excellent source of Vitamin C
- Nutritionally complete treat or sole source diet
- No messy shells to sweep or clog drains
- Nutritionally assayed
- Shelf Life: 6 months, cool dry storage
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Peanuts, Dried Brewers Yeast, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride, Calcium Silicate, Magnesium Stearate, Sodium Propionate, Ascorbic Acid, Kaolin, tBHQ.
Caloric value: 18.1 kcal/Wafer
Nutritional ash content: 6.8%
Nutritional carbohydrate content: 52.0%
Nutritional fat content: 7.5%
Nutritional fibre content: 4.5%
Nutritional moisture content: <10%
Nutritional protein content: 21.5%
Vitamin C: 12.6 mg/Wafer
Vitamin D3: 36.9 IU/Wafer