Order Entry
Puerto Rico
ContactUsLinkComponent
Monkey Dough™ Diet, Bio-Serv
Monkey Dough™ Diet, Bio-Serv
Catalog # 89067-322
Supplier:  Bio-Serv
undefined
Monkey Dough™ Diet, Bio-Serv
Catalog # 89067-322
Supplier:  Bio-Serv
Supplier Number:  F4548

Specifications

  • Description:
    Monkey Dough, Banana Flavor
  • Cat. No.:
    89067-322
  • Packaging:
    3 kg/Pail
  • Supplier no.:
    F4548

Specifications

About this item

Nutritionally complete, extra-soft diet with a cookie dough texture

  • Delicious banana flavor with raisins added
  • Ideal for post-op care and finicky eaters; medications can be added
  • Perfect for geriatrics, weanlings or primates with chewing disabilities
  • Convenient alternative to feeding "soaked biscuits" or making other in-house diets
  • Nutritionally assayed
  • Shelf Life: 6 months frozen; 2 months refrigerated after opening

Ingredients: Whole Milk Powder, Casein, Sucrose, Cellulose, Soybean Oil, Corn Syrup, Raisins, Mineral Mix, Glycerin, Banana Flavor, Sorbic Acid, Ascorbic Acid, Vitamin E Acetate, tBHQ.

Caloric value: 3.64 kcal/g
Nutritional ash content: 3.5%
Nutritional carbohydrate content: 39.5%
Nutritional fat content: 12.0%
Nutritional fibre content: 7.7%
Nutritional moisture content: <13%
Nutritional protein content: 24.6%
Vitamin C: 2.7 mg/g
Vitamin D3: 4.8 IU/g