Mycoplasma Agar, Hardy Diagnostics
10753-452
Product available on GSA Advantage®
:
- Pack type:Plate
- Description:Mycoplasma Agar
- Size:15 x 100 mm
- Environmentally Preferable:
- Cat. No.:10753-452
- Supplier no.:G102
Mycoplasma media is recommended for use in isolation of mycoplasma species associated with bovine mastitis.
- Optically clear polystyrene - For distortion free viewing
- Heavy-Duty- Reduces breakage during shipping
- Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
Polystyrene Petri plates feature raised rings for stacking that prevent sliding.
Colonies have a "fried egg" appearance when viewed at 100X under the microscope.
: Breathable Packaging - Packaged in sleeves of 10 plates in a "breathable" bag that prevents buildup of condensation and excess moisture.