Order Entry
Puerto Rico
ContactUsLinkComponent
Epower™ Saccharomyces kudriavzevii, Microbiologics
Epower™ Saccharomyces kudriavzevii, Microbiologics
Catalog # 89505-746
CAS Number:  
undefined
Epower™ Saccharomyces kudriavzevii, Microbiologics
Catalog # 89505-746
Supplier Number:  0698E3
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Saccharomyces kudriavzevii
  • Storage Conditions:
    Easy storage and no dry ice required, reduces shipping costs
  • Cat. No.:
    89505-746
  • Includes:
    10 Pellets
  • ATCC no.:
    ATCC® 2601™*
  • Supplier no.:
    0698E3

Specifications

About this item

Epower™ QC microorganisms can be used to perform many essential quality control functions including microbial detection and enumeration, equipment calibration, method validation, bioburden determination, and antibacterial effectiveness and lethality testing

Epower™ is packaged in a resealable vial containing 10 pellets of a single quantitative QC microorganism strain. An online certificate of analysis containing documentation of phenotypic, macroscopic and microscopic test results is available.

Available in concentrations ranging from 102 to 108 CFU per pellet, Epower™ lyophilized microorganism preparations are extremely versatile. Epower™ pellets can be used individually or several Epower™ pellets can be combined for a mixed microorganism population challenge. They can also be easily manipulated to deliver a wide variety of concentrations. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.

Certifications: Assured quality, meets USP guidelines.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.