Epower™ Staphylococcus aureus subsp. aureus, Microbiologics
About this item
Epower™ QC microorganisms can be used to perform many essential quality control functions including microbial detection and enumeration, equipment calibration, method validation, bioburden determination, and antibacterial effectiveness and lethality testing
Epower™ is packaged in a resealable vial containing 10 pellets of a single quantitative QC microorganism strain. An online certificate of analysis containing documentation of phenotypic, macroscopic and microscopic test results is available.
Available in concentrations ranging from 102 to 108 CFU per pellet, Epower™ lyophilized microorganism preparations are extremely versatile. Epower™ pellets can be used individually or several Epower™ pellets can be combined for a mixed microorganism population challenge. They can also be easily manipulated to deliver a wide variety of concentrations. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.
Certifications: Assured quality, meets USP guidelines.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
8 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Epower™ QC microorganisms can be used to perform many essential quality control functions including microbial detection and enumeration, equipment calibration, method validation, bioburden determination, and antibacterial effectiveness and lethality testing
Epower™ is packaged in a resealable vial containing 10 pellets of a single quantitative QC microorganism strain. An online certificate of analysis containing documentation of phenotypic, macroscopic and microscopic test results is available.
Available in concentrations ranging from 102 to 108 CFU per pellet, Epower™ lyophilized microorganism preparations are extremely versatile. Epower™ pellets can be used individually or several Epower™ pellets can be combined for a mixed microorganism population challenge. They can also be easily manipulated to deliver a wide variety of concentrations. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.
Certifications: Assured quality, meets USP guidelines.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Documents
Certificates
Search by Lot NumberInstructions for End User Registration with Microbiologics
Additional Product Details
Microbiologics Epower
Product Literature
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 89502-604
- 89503-808
- 89503-812
- 89503-926
- 89503-930
- 89503-934
- 89504-642
- 89504-646
- 89506-456
- Storage Conditions:
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
- Easy storage and no dry ice required, reduces shipping costs
Specifications
Product Family Options
Product Information
- ATCC no.IncludesAvailabilityPrice
- 89502-604Staphylococcus aureus subsp. aureusSupplier Number: 0158E4Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89502-604Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89503-808Staphylococcus aureus subsp. aureusSupplier Number: 0360E3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89503-808Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89503-812Staphylococcus aureus subsp. aureusSupplier Number: 0360E4Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89503-812Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89503-926Staphylococcus epidermidisSupplier Number: 0371E3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89503-926Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89503-930Staphylococcus epidermidisSupplier Number: 0371E4Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89503-930Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89503-934Staphylococcus epidermidisSupplier Number: 0371E8Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89503-934Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89504-642Staphylococcus aureus subsp. aureusSupplier Number: 0485E3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89504-642Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89504-646Staphylococcus aureus subsp. aureusSupplier Number: 0485E7Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89504-646Storage ConditionsEasy storage and no dry ice required, reduces shipping costs - 89506-456Staphylococcus aureus subsp. aureusStaphylococcus aureus subsp. aureusATCC® 6538P™*10 PelletsDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89506-456Storage ConditionsEasy storage and no dry ice required, reduces shipping costs