ContactUsLinkComponent
HIV Serum, Charles River Laboratories Cell Solutions
HIV Serum, Charles River Laboratories Cell Solutions
Catalog # 102830-122
HIV Serum, Charles River Laboratories Cell Solutions
Catalog # 102830-122
Supplier Number:  PB007C-1-HIV
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Serum format:
    Cryopreserved
  • Description:
    HIV Serum
  • Size:
    500 mL
  • Cat. No.:
    102830-122
  • Supplier no.:
    PB007C-1-HIV
Specifications
About this item
Human disease state peripheral blood serum is collected in HemaCare's FDA-registered collection centers or through our hospital or physician office network from IRB consented human volunteer donors
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
Serum is collected from peripheral blood in a plain or Serum Separator Tube.