Order Entry
Puerto Rico
ContactUsLinkComponent
Non-Hodgkins Lymphoma Plasma, Charles River Laboratories Cell Solutions
Non-Hodgkins Lymphoma Plasma, Charles River Laboratories Cell Solutions
Catalog # 102830-142
CAS Number:  
Non-Hodgkins Lymphoma Plasma, Charles River Laboratories Cell Solutions
Catalog # 102830-142
Supplier Number:  PB006C-1-NHL
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Non-Hodgkins Lymphoma Plasma
  • Size:
    2 mL
  • Cat. No.:
    102830-142
  • Format:
    Cryopreserved
  • Supplier no.:
    PB006C-1-NHL

Specifications

About this item

Human disease state peripheral blood plasma is collected in HemaCare's FDA-registered collection centers or through our hospital or physician office network from IRB consented human volunteer donors

Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.

By default, plasma is separated from whole blood which has been collected in a CPD collection bag or EDTA specimen tubes. If possible, HemaCare will utilize any commercially available special collection tubes with anticoagulants per your specifications.