ContactUsLinkComponent
Gastroenterology (GI) Biopsy Collection and Transport Kits, Therapak®
  10497-202
undefined
Gastroenterology (GI) Biopsy Collection and Transport Kits, Therapak®
  10497-202
 :  38873
  • Description:
    GI Biopsy 2-Vial Transport Kit
  • Cat. No.:
    10497-202
  • Supplier no.:
    38873
Both kits feature standardized components to maximize the ease of collecting and packing specimens for transport
Corrugated inserts are printed with the GI biopsy location ID for easy recognition. In addition, a set of jar labels with the location ID is included in the kit to ensure proper placement of the specimen jars at the point of packing and identification once received at the test location. Customization of these kits is available upon request.
Primary receptacles in the form of 20mL jars are 95kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation. The kits come with either two or six 20mL jars pre-filled with 10mL of formalin solution. Jars are secured by a corrugated insert and placed into the provided zip-style biohazard bag with absorbent material for transport.