Eosin Y (yellowish) 5% stain, Sigma-Aldrich®
Catalog # EM-R03040-74
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:500 ml
- Cat. No.:EM-R03040-74
- Supplier no.:R03040-74
Specifications
Specification Test Results
- General composition:Water soluble, aqueous solution.
- Absorbance of solution:Expected maximum absorbance @ 515nm
Specification Test Results
About this item
:
:691.86 g/mol
CAS Number:17372-87-1
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040