ContactUsLinkComponent
Mobilized Peripheral Blood CD34 Depleted Mononuclear Cells (MNC), Charles River Laboratories Cell Solutions
Mobilized Peripheral Blood CD34 Depleted Mononuclear Cells (MNC), Charles River Laboratories Cell Solutions
Catalog # 102830-044
Mobilized Peripheral Blood CD34 Depleted Mononuclear Cells (MNC), Charles River Laboratories Cell Solutions
Catalog # 102830-044
Supplier Number:  M34DC-1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Format:
    Cryopreserved
  • Description:
    Mobilized Peripheral Blood CD34 Depleted Mononuclear Cells
  • Size:
    25 x 10e6 Cells
  • Cat. No.:
    102830-044
  • Supplier no.:
    M34DC-1
Specifications
About this item
Human mobilized peripheral blood CD34 depleted mononuclear cells are isolated from peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic cell separation procedures.
PBMCs are collected from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. Donors are injected with G-CSF (Neupogen® - FDA approved) or Mozobil®, which stimulates the bone marrow to produce a large number of hematopoietic and progenitor stem cells and mobilizes them into the peripheral blood stream. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.