ContactUsLinkComponent
Human Bone Marrow Mesenchymal Stem Cells (MSC), Charles River Laboratories Cell Solutions
Human Bone Marrow Mesenchymal Stem Cells (MSC), Charles River Laboratories Cell Solutions
Catalog # 102830-012
CAS Number:  
Human Bone Marrow Mesenchymal Stem Cells (MSC), Charles River Laboratories Cell Solutions
Catalog # 102830-012
Supplier Number:  BM012C-1
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Bone Marrow Mesenchymal Stem Cells
  • Size:
    1 x 10e6 Cells
  • Cat. No.:
    102830-012
  • Supplier no.:
    BM012C-1
Specifications
About this item
Human bone marrow mesenchymal stem cells (MSC) are isolated by adherence from bone marrow mononuclear cells (MNCs), isolated using a density gradient
Adherent cells were cultured in MEM-Alpha, supplemented with L-Glutamine and Human Platelet Lysate. Human bone marrow is collected from healthy human volunteer donors under IRB approved informed consent via our qualified hospital and physician network.