Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugate ELISA:HRP
- Description:450 nm liquid Nova-Stop solution
- Size:100 ml
- Color Development:Yellow Solute
- Shelf Life:4 years
- Storage Temperature:Room Temperature (25 °C)
- Cat. No.:102867-006
- Supplier no.:NSTP-0100-01
The stop solution provides minimial signal drift compared to sulfuric or hydrochloric acid. Absorbance is read in the yellow spectral range (450 nm) and can be read up to 3 hours after the TMB reaction has been stopped using the Nova-Stop Solution.
- Room Temperature Storage
- 4 Year Shelf Life
- Safe for lab personnel and environment (non-corrosive to both skin and eyes as opposed to the alternative sulfuric acid)
- Ready to use
A liquid stop solution used in end point ELISA applications where a TMB substrate is used as the detection mechanism.