Order Entry
Puerto Rico
ContactUsLinkComponent
BioFX® Chemiluminescent Sensitive Plus HRP Microwell and/or Membrane Substrate, Surmodics IVD
BioFX® Chemiluminescent Sensitive Plus HRP Microwell and/or Membrane Substrate, Surmodics IVD
Catalog # 101396-952
Supplier:  Surmodics
CAS Number:  
BioFX® Chemiluminescent Sensitive Plus HRP Microwell and/or Membrane Substrate, Surmodics IVD
Catalog # 101396-952
Supplier:  Surmodics
Supplier Number:  LERI-0110-2C
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Chemiluminescent Sensitive Plus HRP Microwell and/or Membrane Substrate
  • Size:
    110 ml
  • Enzyme Name:
    Horseradish Peroxidase
  • Shelf Life:
    1.5 years
  • Storage Temperature:
    2 to 8° C
  • Application:
    ELISA, Western Blot, Other Membrane Applications
  • Cat. No.:
    101396-952
  • Supplier no.:
    LERI-0110-2C

Specifications

About this item

Chemiluminescent substrates provide a higher degree of sensitivity than other substrates to enable lower limits of detection to be achieved in ELISA and Western Blot assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers. Surmodics provides multiple 2-component HRP chemiluminescent substrates with various intensities to allow the assay developer to choose the appropropriate substrate for the application.

  • Picogram to femtogram levels of sensitivity
  • High signal-to-noise ratios

BioFX® Chemiluminescent Sensitive Plus HRP Substrate provides detection of peroxidase in ELISA and Western Blot applications.

Certifications: ISO 13485:2016 & 9001:2015 Certified