Order Entry
Puerto Rico
ContactUsLinkComponent
BioFX® DAB Membrane Substrate, Surmodics
BioFX® DAB Membrane Substrate, Surmodics
Catalog # 100359-200
Supplier:  Surmodics
CAS Number:  
BioFX® DAB Membrane Substrate, Surmodics
Catalog # 100359-200
Supplier:  Surmodics
Supplier Number:  DABM-0400-2C
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    DAB Membrane Substrate
  • Size:
    400 ml
  • Storage Conditions:
    2...8 °C
  • Color Development:
    Brown precipitate
  • Enzyme Name:
    Horseradish Peroxidase
  • Shelf Life:
    3 years
  • Application:
    Western Blot
  • Cat. No.:
    100359-200
  • Supplier no.:
    DABM-0400-2C

Specifications

About this item

DAB membrane substrate is supplied as a two component liquid substrate system designed for the rapid detection of peroxidase in membrane applications.

  • Three years shelf life

DAB (3,3’-diaminobenzidine) is oxidized in the presence of peroxidase and hydrogen peroxide resulting in the deposition of a brown alcohol-insoluble precipitate at the site of enzymatic activity. The precipitate may range in color from a light golden brown to dark brown depending upon the amount of enzyme present.

Certifications: ISO 13485:2016 and 9001:2015 certified.