Order Entry
Puerto Rico
ContactUsLinkComponent
BioFX® TMB Enhanced HRP Membrane Substrate
BioFX® TMB Enhanced HRP Membrane Substrate
Supplier:  Surmodics
Certificates
BioFX® TMB Enhanced HRP Membrane Substrate
Supplier:  Surmodics

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugate ELISA:
    HRP
  • Color Development:
    Dark blue precipitate
  • Shelf Life:
    48 months at 4° C or 24 months
    at room temperature
  • Storage Temperature:
    2…25° C
  • Application:
    Western blot, Immunoblot

Specifications