About this item
The one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. The staining pattern provides a dark blue precipitate with low background to increase the signal-to-noise ratios in western blot, dot blots and line assays.
- Intense staining pattern with low backgrounds
- Room temperature stability for 2 years
- Stable at 2-8° C for 4 years
BioFX® TMB One Component HRP Membrane Substrate provides increased sensitivity with low backgrounds.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugate ELISA:HRP
- Color Development:Dark Blue Precipitate
- Shelf Life:48 months at 4° C or 24 months at RT
- Storage Temperature:2...8 °C
- Application:Western Blot, Immunblot