Anti-VGLUT1 Rabbit Polyclonal Antibody
Catalog # 10156-178
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Vesicular glutamate transporter 1
- Antigen Symbol:VGLUT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:150 µL
- Antigen Synonyms:Brain-specific Na(+)-dependent inorganic phosphate cotransporter|VGlU1|Solute carrier family 17 member 7|Vesicular glutamate transporter 1|Bnpi
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:VGLUT1
- Purification:Antigen affinity purification
- Cat. No.:10156-178
- Supplier no.:55491-1-AP
Specifications
About this item
The VGLUT1 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human VGLUT1. This antibody recognizes human antigen. The VGLUT1 antibody has been validated for the following applications: ELISA, IHC, FC analysis.
WB 1 : 600
Type: Primary
Antigen: VGLUT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human