The one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution. Peroxidase reacts with the TMB substrate solution to produce a soluble blue reaction product that can be read at 370 nm or 620-650 nm for ELISA applications. Stop solutions can be added to prevent further color development and reading the absorbance values at the desired wavelengths.
- Prolonged Stability
- Increased Sensitivity
- Negligible Backgrounds
- Ease of manufacturing
BioFX™ TMB Extended Range HRP Microwell Substrate is a low-kinetic range substrate for ELISA applications.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugate ELISA:HRP
- Kinetic Rate:Slowest
- Color Development:Yellow/Blue Solute
- Shelf Life:2.5 years
- Storage Temperature:2...8 °C