Anti-MECR Rabbit Polyclonal Antibody
Catalog # 10155-792
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Mitochondrial Trans-2-Enoyl-CoA Reductase
- Antigen Symbol:MECR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:FASN2B|Trans-2-enoyl-CoA reductase|NBRF1|HsNrbf-1|CGI-63|mitochondrial|NRBF1|Nuclear receptor-binding factor 1
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:Mecr
- Purification:Protein A purification
- Cat. No.:10155-792
- Supplier no.:51025-2-IG
Specifications
About this item
The Mecr antibody from Proteintech is a rabbit polyclonal antibody to a peptide of mouse Mecr. This antibody recognizes human, mouse antigen. The Mecr antibody has been validated for the following applications: ELISA, WB, IHC analysis.
WB 1 : 600
Type: Primary
Antigen: MECR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse