Anti-LRRC18 Rabbit Polyclonal Antibody
Catalog # 10155-242
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Leucine Rich Repeat Containing 18
- Antigen Symbol:LRRC18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:LRRC18
- Purification:Antigen affinity purification
- Cat. No.:10155-242
- Supplier no.:24571-1-AP
Specifications
About this item
The LRRC18 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LRRC18. This antibody recognizes human, mouse antigen. The LRRC18 antibody has been validated for the following applications: ELISA, WB, IHC, IP analysis.
WB 1 : 2000
Type: Primary
Antigen: LRRC18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse