Order Entry
Puerto Rico
ContactUsLinkComponent
1-Bromo-3-methoxypropane for synthesis, Sigma-Aldrich®
Flammable
Toxic
1-Bromo-3-methoxypropane for synthesis, Sigma-Aldrich®
Catalog # EM8.43834.0025
Supplier:  MilliporeSigma
CAS Number:  
Flammable
Toxic
1-Bromo-3-methoxypropane for synthesis, Sigma-Aldrich®
Catalog # EM8.43834.0025
Supplier:  MilliporeSigma
Supplier Number:  8.43834.0025
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass Bottle
  • Size:
    25 ml
  • Cat. No.:
    EM8.43834.0025
  • Supplier no.:
    8.43834.0025

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    153.02 g/mol
  • Boiling Point:
    121 °C (1013 hPa)
  • Vapor Pressure:
    6.6 hPa (20 °C)
  • Flash Point:
    43 °C
  • Density:
    1.35 g/cm3
  • pH:
    6 - 7 (H2O)
  • Hill Formula:
    C4H9BrO

Specification Test Results

About this item

 :
 :153.02 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ200
UN:1993
 :GHS02
 :GHS06
CAS Number:36865-41-5
Formula:C₄H₉BrO
MDL Number:MFCD02258473