

Allyl acetate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:591-87-7
:
Formula:C₅H₈O₂
MDL Number:MFCD00008721
:100.12 g/mol
ADR:3 (6.1),II
:M-Clarity™ quality level = MQ200
UN:2333
:GHS02
:GHS06
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Solubility:28 g/l (20 °C)
- Melting Point:-96 °C
- Molar Mass:100.11 g/mol
- Boiling Point:103 - 104 °C (1013 mbar)
- Vapor Pressure:27.2 hPa (20 °C)
- Flash Point:6 °C
- Refractive Index:1.4042 (20 °C, 589 nm)
- Explosion Limit:2.1 - 13.0 %(V)
- Density:0.93 g/cm3 (20 °C)
- Ignition Point:374 °C
- Synonyms:Acetic acid allyl ester
- Hill Formula:C5H8O2
- Chemical Formula:CH3COOCH2CH=CH2
Specification Test Results
Frequently Bought Together