Order Entry
Puerto Rico
ContactUsLinkComponent
D-(-)-Diethyl tartrate for synthesis, Sigma-Aldrich®
D-(-)-Diethyl tartrate for synthesis, Sigma-Aldrich®
Catalog # EM8.18685.0025
Supplier:  MilliporeSigma
CAS Number:  
D-(-)-Diethyl tartrate for synthesis, Sigma-Aldrich®
Catalog # EM8.18685.0025
Supplier:  MilliporeSigma
Supplier Number:  8.18685.0025
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    25 ml
  • Cat. No.:
    EM8.18685.0025
  • Supplier no.:
    8.18685.0025

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    17 °C
  • Molar Mass:
    206.19 g/mol
  • Boiling Point:
    162 °C (25 hPa)
  • Vapor Pressure:
    93 °C
  • Flash Point:
    1.4467 (20 °C, 589 nm)
  • Density:
    1.20 g/cm3 (20 °C)
  • Synonyms:
    L-(+)-Tartaric acid diethyl ester, DET, (-)-Diethyl-D-tartrate
  • Hill Formula:
    C8H14O6
  • Chemical Formula:
    C2H5OOC(CHOH)2COOC2H5

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :206.2 g/mol
CAS Number:13811-71-7
Formula:C₈H₁₄O₆
MDL Number:MFCD00064451