Order Entry
Puerto Rico
ContactUsLinkComponent
N,N-Dimethylglycine hydrochloride for synthesis, Sigma-Aldrich®
N,N-Dimethylglycine hydrochloride for synthesis, Sigma-Aldrich®
Catalog # EM8.18448.0010
Supplier:  MilliporeSigma
CAS Number:  
N,N-Dimethylglycine hydrochloride for synthesis, Sigma-Aldrich®
Catalog # EM8.18448.0010
Supplier:  MilliporeSigma
Supplier Number:  8.18448.0010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, pre-packed
  • Size:
    10 g
  • Cat. No.:
    EM8.18448.0010
  • Supplier no.:
    8.18448.0010

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    190 - 193 °C
  • Molar Mass:
    139.58 g/mol
  • Synonyms:
    N,N-Dimethylaminoacetic acid hydrochloride
  • Hill Formula:
    C4H10ClNO2
  • Chemical Formula:
    (CH3)2NCH2COOH * HCl

Specification Test Results

About this item

 :
 :139.58 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:2491-06-7
Formula:C₄H₁₀ClNO₂
MDL Number:MFCD00012610