Order Entry
Puerto Rico
ContactUsLinkComponent
2,4-Dihydroxy-3,6-dimethylbenzoic acid for synthesis, Sigma-Aldrich®
2,4-Dihydroxy-3,6-dimethylbenzoic acid for synthesis, Sigma-Aldrich®
Catalog # EM8.14703.0005
Supplier:  MilliporeSigma
CAS Number:  
2,4-Dihydroxy-3,6-dimethylbenzoic acid for synthesis, Sigma-Aldrich®
Catalog # EM8.14703.0005
Supplier:  MilliporeSigma
Supplier Number:  8.14703.0005
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 g
  • Cat. No.:
    EM8.14703.0005
  • Supplier no.:
    8.14703.0005

Specifications

Specification Test Results

  • Melting Point:
    187 - 189 °C
  • Molar Mass:
    182.17 g/mol
  • Synonyms:
    3,6-Dimethyl-ß-resorcylic acid
  • Hill Formula:
    C9H10O4

Specification Test Results

About this item

 :
 :182.18
 :M-Clarity™ quality level = MQ200
CAS Number:4707-46-4
Formula:C₉H₁₀O₄
MDL Number:MFCD00239340