Order Entry
Puerto Rico
ContactUsLinkComponent
Palladium(II) nitrate dihydrate Pd 40% for synthesis, Sigma-Aldrich®
Oxidizing
Toxic
Corrosive
Environment
Palladium(II) nitrate dihydrate Pd 40% for synthesis, Sigma-Aldrich®
Catalog # EM8.14573.0001
Supplier:  MilliporeSigma
CAS Number:  
Oxidizing
Toxic
Corrosive
Environment
Palladium(II) nitrate dihydrate Pd 40% for synthesis, Sigma-Aldrich®
Catalog # EM8.14573.0001
Supplier:  MilliporeSigma
Supplier Number:  8.14573.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, pre-packed
  • Size:
    1 g
  • Cat. No.:
    EM8.14573.0001
  • Supplier no.:
    8.14573.0001

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (slow decomposition)
  • Molar Mass:
    266.44 g/mol
  • pH:
    1 (1 g/l, H2O, 20 °C)
  • Hill Formula:
    N2O6Pd * 2 H2O
  • Chemical Formula:
    Pd(NO3)2 * 2 H2O

Specification Test Results

About this item

 :
 :266.46
ADR:5.1 (8),II
 :M-Clarity™ quality level = MQ200
UN:3085
 :GHS03
 :GHS06
 :GHS05
 :GHS09
CAS Number:32916-07-7
Formula:N₂O₆Pd·2H₂O