

Aminoguanidinium hydrogen carbonate for synthesis, Sigma-Aldrich®
Catalog # EM8.01417.0250
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:250 g
- Cat. No.:EM8.01417.0250
- Supplier no.:8.01417.0250
Specifications
Specification Test Results
- Solubility:< - 5 g/l (20 °C)
- Melting Point:125 °C decomposes
- Molar Mass:136.11 g/mol
- Bulk Density:700 kg/m3
- Density:1.56 g/cm3 (20 °C)
- pH:8.9 (5 g/l, H2O, 20 °C)
- Ignition Point:245 °C
- Synonyms:Aminoguanidine bicarbonate
- Hill Formula:C2H8N4O3
- Chemical Formula:CH6N4 * H2CO3
Specification Test Results
About this item
:
ADR:9,III
:M-Clarity™ quality level = MQ200
:136.11 g/mol
UN:3077
:GHS07
:GHS09
CAS Number:2582-30-1
Formula:C₂H₈N₄O₃
MDL Number:MFCD00012949