Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-HSP90 Mouse Monoclonal Antibody [clone: 3F11C1]
Anti-HSP90 Mouse Monoclonal Antibody [clone: 3F11C1]
Catalog # 10155-906
Supplier:  Proteintech
undefined
Anti-HSP90 Mouse Monoclonal Antibody [clone: 3F11C1]
Catalog # 10155-906
Supplier:  Proteintech
Supplier Number:  60318-1-IG-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    HSP90
  • Clonality:
    Monoclonal
  • Clone:
    3F11C1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Storage Temperature:
    Store at –20 °C
  • Immunogen:
    HSP90
  • Purification:
    Protein A purification
  • Cat. No.:
    10155-906
  • Supplier no.:
    60318-1-IG

Specifications

About this item

The HSP90 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human HSP90. This antibody recognizes human antigen. The HSP90 antibody has been validated for the following applications: ELISA, WB, FC analysis.

IHC 1 : 50 ; WB 1 : 300

Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone: 3F11C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human