Anti-LZTS2 Rabbit Polyclonal Antibody
150 µL
10156-244
: 15677-1-AP-150UL
The LZTS2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human LZTS2. This antibody recognizes human,mouse,rat antigen. The LZTS2 antibody has been validated for the following applications: ELISA, IHC, WB analysis.
WB 1 : 600 ; IHC 1 : 50
Type: Primary
Antigen: LZTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Size:150 µL
- Clonality:Polyclonal
- Purification:Antigen affinity purification
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:10156-244
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:LZTS2
- Immunogen:LZTS2
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:Protein LAPSER1|hLZTS2|LAPSER1|Leucine zipper putative tumor suppressor 2|LZTS2|KIAA1813
- Antigen Name:Leucine Zipper Putative Tumor Suppressor 2
- ELISA:Yes
- Storage Temperature:Store at –20 °C



