Anti-GDAP2 Rabbit Polyclonal Antibody
Catalog # 10156-252
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Ganglioside induced differentiation associated Protein 2
- Antigen Symbol:GDAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:MACROD3|GDAP2_HUMAN
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:GDAP2
- Purification:Antigen affinity purification
- Cat. No.:10156-252
- Supplier no.:17246-1-AP
Specifications
About this item
The GDAP2 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human GDAP2. This Antibody recognizes human,mouse,rat Antigen. The GDAP2 Antibody has been validated for the following applications: ELISA, WB analysis.
WB 1 : 300 ; IHC 1 : 50
Type: Primary
Antigen: GDAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat