Anti-ACTR1B Rabbit Polyclonal Antibody
10154-424
- Antibody Type:Primary
- Antigen Name:Actin-Related Protein 1B
- Antigen Symbol:ACTR1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:centractin beta|PC3|yeast) homolog B (centractin beta)|yeast homolog B|ARP1B|ARP1 actin-related protein 1 homolog B|CTRN2|ARP1|ARP1 (actin-related protein 1|centractin beta (yeast)
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:ACTR1B
- Purification:Antigen affinity purification
- Cat. No.:10154-424
- Supplier no.:11023-1-AP
The ACTR1B antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human ACTR1B. This antibody recognizes human,mouse,rat antigen. The ACTR1B antibody has been validated for the following applications: ELISA, IHC, IF, WB analysis.
WB 1 : 1200 ; IHC 1 : 100 ; IF 1 : 25
Type: Primary
Antigen: ACTR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat