Anti-TRPS1 Rabbit Polyclonal Antibody
Catalog # 10154-612
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Trichorhinophalangeal Syndrome I
- Antigen Symbol:TRPS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:150 µL
- Antigen Synonyms:trichorhinophalangeal syndrome I|GC79|LGCR
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:TRPT1
- Purification:Antigen affinity purification
- Cat. No.:10154-612
- Supplier no.:15541-1-AP
Specifications
About this item
The TRPT1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TRPT1. This antibody recognizes human,mouse,rat antigen. The TRPT1 antibody has been validated for the following applications: ELISA, IHC analysis.
WB 1 : 2000 ; IF 1 : 25
Type: Primary
Antigen: TRPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat