To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Myosin Heavy Chain 7
- Antigen Symbol:MYH7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:SPMD|CMH1|SPMM|MYHCB|MPD1|CMD1S
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:MYH7-specific
- Purification:Antigen affinity purification
- Cat. No.:10155-048
- Supplier no.:22280-1-AP
Specifications
About this item
The MYH7-specific antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human MYH7-specific. This antibody recognizes human, mouse antigen. The MYH7-specific antibody has been validated for the following applications: ELISA, IHC, WB, IP analysis.
WB 1 : 300
Type: Primary
Antigen: MYH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse