Anti-FBXL15 Rabbit Polyclonal Antibody
Catalog # 10154-902
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:F-Box and Leucine-rich Repeat Protein 15
- Antigen Symbol:FBXL15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:Fbl15|PSD|JET|FBXO37
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:FBXL15
- Purification:Antigen affinity purification
- Cat. No.:10154-902
- Supplier no.:20895-1-AP
Specifications
About this item
The FBXL15 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FBXL15. This antibody recognizes human antigen. The FBXL15 antibody has been validated for the following applications: ELISA, WB analysis.
WB 1 : 600 ; IF 1 : 50 ; IHC 1 : 50
Type: Primary
Antigen: FBXL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human