Anti-P2RY13 Rabbit Polyclonal Antibody
Catalog # 10154-874
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Purinergic Receptor P2Y, G-Protein Coupled 13
- Antigen Symbol:P2RY13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:SP174|FKSG77|GPR94|GPCR1|GPR86|P2Y13
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:P2RY13
- Purification:Antigen affinity purification
- Cat. No.:10154-874
- Supplier no.:20335-1-AP
Specifications
About this item
The P2RY13 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human P2RY13. This antibody recognizes human, mouse antigen. The P2RY13 antibody has been validated for the following applications: ELISA, WB, IHC analysis.
WB 1 : 1000 ; IHC 1 : 50
Type: Primary
Antigen: P2RY13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse