About this item
StabilZyme® AP and StabilZyme® Protein-Free AP stabilizers provide assay developers a complete set of tools to stabilize different AP conjugates for long term, in-solution stability.
- Protein-free formulation
- Extended shelf life of kits
StabilZyme® Protein-Free AP Stabilizer eliminates interference and cross-reactivity caused by proteins, delivering optimal performance for even the most sensitive immunoassays.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein:Protein Free
- Shelf Life:2.5 years
- Storage Buffer:MOPS
- Storage Temperature:2 to 8 °C or Room Temperature
- pH:6.3-7.3