ContactUsLinkComponent
MIDORI Green Advance Nucleic Acid Staining Solution, Bulldog-bio
MIDORI Green Advance Nucleic Acid Staining Solution, Bulldog-bio
Catalog # 102407-968
Supplier:  Bulldog Bio
CAS Number:  
undefined
MIDORI Green Advance Nucleic Acid Staining Solution, Bulldog-bio
Catalog # 102407-968
Supplier:  Bulldog Bio
Supplier Number:  MG04
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    MIDORI Green Advance
  • Size:
    1 ml
  • Cat. No.:
    102407-968
  • Supplier no.:
    MG04
Specifications
About this item
Midori Green Advance — Safe DNA Stain, a safe alternative to ethidium bromide
  • Compatible with UV transilluminators
  • Compatible with blue LED illuminators
MIDORI Green Advance can stain single- and double-stranded nucleic acids with sensitivity similar to that of ethidum bromide(EtBr). Like EtBr, Midori Green Advance is added directly to molten agarose during the gel pouring step. This makes it a no-brainer as a drop-in replacement for your lab’s current protocols. Not only can you use your existing UV transilluminator and filter sets — MIDORI Green Advance works best when matched with blue LED-, or in particular, Blue/Green LED-based illuminators and gel documentation systems. These environmentally-friendly light tables are the perfect complement for any MIDORI Green stain, as you can work shield-less and goggle-less when excising bands or imaging migration. And because blue or green light does not cross-link DNA, you never need to worry about damaging DNA during cloning and photographing sessions.
MIDORI Green Advance shows a very high sensitivity even for small DNA fragments. The dilution factor of MIDORI Green Advance can be as high as 1:25000. Hence, 4 μL to 6 μL are enough for a 100 mL agarose gel, resulting in about 17 to 25 liters of stained agarose gels per 1 mL tube of MIDORI Green Advance.