Order Entry
Puerto Rico
ContactUsLinkComponent
TruLab 1320 Dual Channel Laboratory Instrument for pH, mV (ORP) and Temperature With Printer, YSI
TruLab 1320 Dual Channel Laboratory Instrument for pH, mV (ORP) and Temperature With Printer, YSI
Catalog # 10122-578
Supplier:  YSI
undefined
TruLab 1320 Dual Channel Laboratory Instrument for pH, mV (ORP) and Temperature With Printer, YSI
Catalog # 10122-578
Supplier:  YSI
Supplier Number:  1GA340PY
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • No. of Channels:
    2 (measurement)
  • Temperature Range:
    –5 to 105°C (23.0 to 221°F)
  • Description:
    Meter only
  • Temperature Accuracy:
    (+1 digit): ±0.1
  • Temperature Resolution:
    0.1
  • Storage Temperature:
    –25 to 65°C (–13 to 149°F)
  • Parameter:
    pH, ORP (mV), Ammonia, Ammonium, Bromide, Cadmium, Calcium, Chloride, Copper, Cyanide, Fluoride, Iodide, Lead, Nitrate, Potassium, Silver/Sulfide, Sodium, Temperature
  • Calibration Points:
    pH 1, 2, 3, 4 or 5 point with or without autobuffer recognition; USA and NIST and 22 pre–programmed buffer sets. ISE 2 to 7 points
  • Data Logging Capacity:
    5,000 data sets auto logging; 500 data sets manual logging
  • Display:
    Backlit graphic display
  • ISE Resolution:
    0.001; 0.01; 0.1; 1 (mg/l, mg/kg, mmol/l, ppm, %) (µmol/l)
  • ISE Range:
    0.000 to 9.999; 10.00 to 99.99; 100.0 to 999.9; 1000 to 999999; 0.000 to 9.999; 10.00 to 99.99; 100.0 to 999.9; 1000 to 9999 (mg/l, mg/kg, mmol/l, ppm, %) (µmol/l)
  • Power supply:
    4 alkaline AA batteries fully charged or included universal power supply; approximately 150 hours with illumination or 1000 without
  • Temperature Compensation:
    Automatic or Manual; Manual Input -25 to 130°C (-13 to 266°F) in increments of 1
  • Weight:
    Model 1320 = 0.8 kg (1.8 lbs.); Model 1320P = 1.0 kg (2.0 lbs.)
  • mV accuracy:
    (+1 digit): ±0.3; ±1.0 (temp of test sample 15 to 35°C)
  • mV range:
    (+1 digit): ±0.3; ±1.0 (temp of test sample 15 to 35°C)
  • mV resolution:
    (+1 digit): ±0.3; ±1.0 (temp of test sample 15 to 35°C)
  • pH Accuracy:
    (+1 digit): ±0.1; ±0.01; ±0.005 (temp of test sample 15 to 35°C; within a range of ±2 pH of the calibration point)
  • pH Range:
    –2.0 to 20.0; –2.00 to 20.00; –2.000 to 19.999
  • pH Resolution:
    0.1; 0.01; 0.001
  • Cat. No.:
    10122-578
  • Supplier no.:
    1GA340PY

Specifications

About this item

The YSI TruLab® line includes the 1110 (single channel), 1310 and 1310P (single channel) and 1320 and 1320P (dual channel) instruments providing easy-to-use and calibrate instruments ideal for the laboratory.

Measure pH, mV (ORP) and temperature accurately in the laboratory with the YSI pH 1320. This benchtop instrument for basic, routine measurements in the laboratory is an accurate and easy-to-use solution for repeatable pH or mV measurements.

Convenient/Reliable–An adjustable timer indicates when to perform the next calibration routine, improving measurement accuracy.

ISE measurements include Blank value correction, increment methods: Known addition (single); Known subtraction; Sample addition; Sample subtraction; Known addition (double).

Certifications: CE, cETLus, electrical protective class III, IP43.