Dropper® Urine Dipstick Controls, Quantimetrix
82024-128
:
- Description:Dropper® Urine Dipstick Control
- Cat. No.:82024-128
- Volume:4× 25 ml
- Includes:Level 1 and 2 Set
- Supplier no.:1440-02
Dropper Urine Dipstick Control, monitors the performance of visual and instrument readings of urine dipsticks by dropping the control directly onto the dipstick. Features an easy-to-use dropper bottle for simple dispensing and maximum use. We include values for all dipstick analytes plus microalbumin and creatinine – and qualitative results for hCG early pregnancy detection test methods. Dropper Urine Dipstick Control is designed for use with most urinalysis reagent strips. It can also be used for confirmatory tests and refractometry.
- Easy-to-use dropper bottles
- Liquid, made with human urine
- 18-month shelf life and open vial stability from date of manufacture
- when stored at 2 to 8°C
: Box includes 2 vials each of Level 1 and 2. Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.