Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Detector Mono™, Immunostics

Detector Mono™, Immunostics

Detector Mono™, 25
Catalog # 10147-072
Supplier:   IMMUNOSTICS, INC.
Supplier #:  IDM-25
Detector Mono™, Immunostics
Catalog # 10147-072
Supplier:  IMMUNOSTICS, INC.
CAS Number:  
Detector Mono™, Immunostics
Detector Mono™, Immunostics

Detector Mono™, 25

CAS Number:  
Price
$208.30
Log in for availability
Kit of 25
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Detector Mono™, Immunoassay for the qualitiative detection of infectious mononucleosis heterophile antibodies as well as EBV viral proteins associated with infectious Mononucleosis antibodies in whole blood, plasma, and serum.

  • Sensitivity 99.9%
  • Specificity 98.8%
  • Test time 8 Minutes

Certifications: CLIA Waived (For Whole Blood Only). CE Marked.

Ordering information: Kit includes 25 Test Cassettes, 25 Disposable Pipettes, 1 Vial of Developing Buffer, 1 Vial of Positive Control, 1 Vial of Negative Control, and 1 Product Insert.

Documents

Specifications

Specifications

  • Description:
    Detector Mono™
  • No. of tests:
    25

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.