ContactUsLinkComponent
hema-screen STAT, Immunostics
hema-screen STAT, Immunostics
Catalog # 10147-018
Supplier:  IMMUNOSTICS, INC.
CAS Number:  
undefined
hema-screen STAT, Immunostics
Catalog # 10147-018
Supplier:  IMMUNOSTICS, INC.
Supplier Number:  HSTAT-50
CAS Number:  
Specifications
  • Description:
    hema-screen STAT Slides with On Slide Developer
  • Cat. No.:
    10147-018
  • Supplier no.:
    HSTAT-50
  • No. of tests:
    50
Specifications
About this item
A guaiac-based in vitro slide method for the qualitative detection of fecal occult blood with on-slide developer.
Certifications: CLIA Waived. CE Marked.