Small Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
Catalog # 89007-222
Supplier: Brady Worldwide
Specifications
- Color:Red
- Description:Small Universal Valve Lockout Base Clamping Unit
- Material:Industrial-Grade Steel and Nylon
- Cat. No.:89007-222
- Supplier no.:50924
Specifications
About this item
The universal valve lockouts are effective for all types of standard valves.
- Versatile
- Rugged
- Easy-to-use
- Lightweight
They can also be used to lock out large levers, T-handles, and hard to secure mechanical devices. New open-ended clamp fits over closed rings and wide handles. Constructed of industrial-grade steel and nylon for extra impact and chemical resistance.
Ordering information: Base unit only (cable and blocking arm sold separately).