ContactUsLinkComponent
Large Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
Large Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
  89007-220
undefined
Large Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
  89007-220
 :  50899
  • Color:
    Red
  • Material:
    Industrial-Grade Steel and Nylon
  • Cat. No.:
    89007-220
  • Supplier no.:
    50899
Universal valve lockouts are effective for all types of standard valves
  • Versatile and rugged
  • lightweight and easy-to-use
New open-ended clamp fits over closed rings and wide handles. Constructed of industrial-grade steel and nylon for extra impact and chemical resistance.
They can also be used to lock out large levers, T-handles, and hard-to-secure mechanical devices.
 : Cable and blocking arm sold separately.