All-Purpose Cable Lockout, with 8' Sheathed Cable, Brady Worldwide®
10154-258
- Color:Red
- Description:All-Purpose Cable Lockout, with 8' Sheathed Cable
- Material:Impact Modified Glass Filled Nylon
- Size:8' L x 3/16" Dia.
- Cat. No.:10154-258
- Supplier no.:50943
Cable lockouts are incredibly versatile, making them perfect for any unusual energy isolating devices that are difficult to lock out
Compact, lightweight body is made of rugged, impact modified glass filled nylon giving them superior chemical, corrosion, and temperature resistant properties. Sheathed metal cable ha as 3/16" diameter, including vinyl coating. Ergonomic design makes it a "cinch" to tighten the cable; simply hold device in one hand and cinch tight with the other. Built-in cable loop keeps cable neatly coiled when stored and prevents it from dangling loose when device is in use. The four lock holes in handle accommodate most lock shackles and hasps. Additional 8', 12', 16', and 20' cables are sold separately.