ContactUsLinkComponent
Large Enclosed Lockout Station (Station Only), Brady Worldwide®
Large Enclosed Lockout Station (Station Only), Brady Worldwide®
Catalog # 89007-242
Supplier:  Brady Worldwide
undefined
Large Enclosed Lockout Station (Station Only), Brady Worldwide®
Catalog # 89007-242
Supplier:  Brady Worldwide
Supplier Number:  50996
Specifications
  • Color:
    Red on White
  • Description:
    Large Enclosed Lockout Station (Station Only)
  • Material:
    Polystyrene
  • Size:
    26"H x 19.5"W x 5"D
  • Cat. No.:
    89007-242
  • Supplier no.:
    50996
Specifications
About this item
The large enclosed lockout station with components provides everything needed for locking out all types of valves and plugs
Constructed of sturdy white polystyrene with predrilled holes for easy wall mounting. Pegboard-style holes for hooks are drilled into the back (6 hooks supplied standard). Built-in lock rail holds 15 to 20 locks, depending on lock size. Station is part of Large Lockout Station Kits 99698, 99699, and 99697.