Order Entry
Puerto Rico
ContactUsLinkComponent
MicroPulser™ Electroporator, Bio-Rad
MicroPulser™ Electroporator, Bio-Rad
Catalog # 100446-902
Supplier:  Bio-Rad
MicroPulser™ Electroporator, Bio-Rad
Catalog # 100446-902
Supplier:  Bio-Rad
Supplier Number:  1652100

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    MicroPulser™ Electroporator
  • W×D×H:
    29×21×8 cm
  • Ambient Operating Temperature:
    3.5...35 °C
  • Ambient Operating Humidity:
    0 - 95% (noncondensing)
  • Output:
    3,000V Peak into > 600 W Load, Limited at 100 A Peak Maximum
  • Input:
    100–120 V or 220–240 V
  • Protocol:
    5 Bacterial, 5 Fungal
  • Waveform:
    Decaying or truncated decaying exponential-decay with resistor capacitor time constant
  • Weight:
    2.9 kg
  • Cat. No.:
    100446-902
  • Supplier no.:
    1652100

Specifications

About this item

The MicroPulser™ Electroporator provides safe and reproducible transformation of bacteria, yeast, and other microorganisms

  • One-button pulse delivery
  • Rapid charge time for fast sample handling
  • Arc quenching (ARQ) system significantly reduces arcing, protecting against sample loss
  • High-voltage 3,000 V capability improves efficiency in larger-volume cuvettes
  • Compact, space-saving design
  • Audible and visible pulse indicators
  • Time constant and actual volts displayed to monitor reproducibility

Manual programming allows voltage to be selected in a 200–3,000 V range, with 10 V precision, and allows pulse width to be selected in a 1.0–4.0 ms range with 0.1 ms precision.

DNA, RNA, siRNA, proteins, virions, nucleotides, carbohydrates, and dyes can be electroporated into cells. The electroporator has preset programs optimized for common bacteria and fungi, and the ability to manually program voltage and pulse width.

Certifications: Meets EN61000- 3-2 and EN61000-3-3 harmonic, flicker, and voltage fluctuation standards, FCC, Class A IEC 1010, C.