Mouse IgG1 Monoclonal Isotype Control [clone: MOPC-21] (violetFluor® 450)
10208-508
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Conjugation:VioletFluor® 450
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Size:100 µg
- Storage Temperature:2-8°C protected from light
- Tested Applications:Flow Cytometry
- Cat. No.:10208-508
- Supplier no.:75-4714-U100
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
Type: Control
Antigen: MOPC21
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: