Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD27 Mouse Monoclonal Antibody (violetFluor® 450) [clone: O323]
Anti-CD27 Mouse Monoclonal Antibody (violetFluor® 450) [clone: O323]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD27 Mouse Monoclonal Antibody (violetFluor® 450) [clone: O323]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10208-506
  • 10208-504
  • Antigen Symbol:
  • CD27
  • CD27
  • Antigen Name:
  • CD27 molecule
  • CD27 molecule
  • Antigen Synonyms:
  • Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
  • Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • VioletFluor® 450
  • VioletFluor® 450
  • Clone:
  • O323
  • O323
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Human
  • Baboon, Cynomolgus monkey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 939
  • 939
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Storage Temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications